missing translation for 'onlineSavingsMsg'
Learn More

Cobamamide, TRC

Product Code. 30134723
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134723 1 g 1g
17978104 10 mg 10mg
17908584 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134723 Supplier Toronto Research Chemicals Supplier No. TRCC6331301G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13870-90-1
Molecular Formula C72 H100 Co N18 O17 P
Molecular Weight (g/mol) 1579.58
Synonym Cobinamide, Co-(5'-deoxyadenosin-5'-yl)-, f-(dihydrogen phosphate), inner salt, 3'-ester with (5,6-dimethyl-1-α-D-ribofuranosyl-1H-benzimidazole-κN3) (9CI), Cobinamide, Co-(5'-deoxyadenosine-5') deriv., hydroxide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-α-D-ribofuranosyl-1H-benzimidazole, Cobinamide, Co-(5'-deoxyadenosine-5') deriv., hydroxide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-α-D-ribofuranosylbenzimidazole (8CI), (5,6-dimethylbenzimidazolyl)cobamide coenzyme, (5'-Deoxy-5'-adenosyl)cobamide coenzyme, 5,6-Dimethylbenzimidazolyl-Co-5'-deoxyadenosylcobamide, 5-Deoxyadenosylcobalamin, 5'-Deoxy-5'-adenosylcobalamin, 5'-Deoxyadenosyl vitamin B12, 5'-Deoxyadenosyl-5,6-dimethylbenzimidazolylcobamide, 5'-Deoxyadenosylcobalamin, Actimide, Adenosyl B12, Adenosylcobalamin, Anabasi, Betarin, Calomide, Cobalamin coenzyme, Cobalamin, Co-(5'-deoxy-5'-adenosyl)-, Cobalion, Cobamamide, Cobamide coenzyme, (5,6-dimethyl-1H-benzimidazolyl)-, Cobamide coenzyme, (5'-deoxy-5'-adenosyl)-, Cobamide, Co-(5'-deoxy-5'-adenosyl)-α-(5,6-dimethyl-1H-benzimidazolyl)-, Cobanzyme, Cobatlamin S, Cobazymase, Coenzile, Coenzyme B12, DBC, DBC coenzyme, DBCC, Deoxyadenosylcobalamin, Dibencozide, Dibenzcozamide, Dimebenzcozamide, Dolonevran, Enzicoba, Funacomide, Heraclene, Hi-Fresmin, Hycobal, Indusil, LM 176, Represil, Sabalamin, Vitamin B12 coenzyme, Vitamin B12, Co-(5'-deoxy-5'-adenosyl) deriv., Xobaline, α-(5,6-Dimethylbenzimidazolyl)-Co-5'-deoxyadenosylcobamide, α-(5,6-Dimethylbenzimidazolyl)cobamide coenzyme
IUPAC Name (2S,3S,4R)-2-(6-aminopurin-9-yl)-5-methanidyloxolane-3,4-diol;cobalt(3+);[(2R,5S)-5-(5,6-dimethylbenzimidazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl] 1-[3-[(2R,3R,4Z,7S,9Z,12S,13S,14Z,17S,18S,19R)-2,13,18-tris(2-amino-2-oxoethyl)-7,12,17-tris(3-amino-3-oxopropyl)-3,5,8,8,13,15,18,19-octamethyl-2,7,12,17-tetrahydro-1H-corrin-21-id-3-yl]propanoylamino]propan-2-yl phosphate
SMILES C[C@@H]1CNC(=O)CC[C@]2(C)[C@@H](CC(=O)N)[C@H]3N4C2=C(C)C5=N6C(=CC7=N8C(=C(C)C9=N([C@]3(C)[C@@](C)(CC(=O)N)[C@@H]9CCC(=O)N)[Co+]468(C[C@H]%10O[C@H]([C@H](O)[C@@H]%10O)n%11cnc%12c(N)ncnc%11%12)N%13=CN([C@H]%14O[C@H](CO)[C@@H](OP(=O)([O-])O1)[C@H]%14O)c%15cc(C)c(C)cc%13%15)[C@@](C)(CC(=O)N)[C@@H]7CCC(=O)N)C(C)(C)[C@@H]5CCC(=O)N

Specifications

Quantity 1 g
InChI Formula InChI=1S/C62H90N13O14P.C10H12N5O3.Co/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;1-4-6(16)7(17)10(18-4)15-3-14-5-8(11)12-2-13-9(5)15;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);2-4,6-7,10,16-17H,1H2,(H2,11,12,13);/q;;+2/p-2/t31-,34-,35-,36-,37+,41-,52-,53-,56-,57+,59-,60+,61+,62+;4-,6-,7-,10-;/m11./s1
Formula Weight 1578.6583
Percent Purity >95%
Chemical Name or Material Adenosylcobalamine (Coenzyme B12)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.