Learn More
KARTELL COBAS MIRA™ Multi-Curvette
Product Code. 10767524
Change view
Click to view available options
Quantity:
1 Segment
15 Segments
Unit Size:
Pack of 450
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10767524 1 Segment Pack of 450
10551393 15 Segments Pack of 450
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10767524 Supplier KARTELL Supplier No. 0150100
  • ONLINE EXCLUSIVE PRICE
    374.00 €
    298.42 € / Pack of 450
    Save 75.58 €
    20% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The COBAS MIRA™ Multi-Curvette is a 12-position polymethyl methacrylate (PMMA) multiple curvette segement for the Cobas Mira™ analyzer.
  • 12-position polymethyl methacrylate (PMMA) multiple cuvette segments for the Cobas Mira™; analyzer.
  • A high-tech production process and stringent quality control guarantee high reproducibility.
  • Available in racks of 15 cuvette segments for devices with continuous throughput or in single segments.
Specifications
Type Multi-cuvette
Sample Type Serum, Plasma, Urine, Whole Blood
Quantity 1 Segment