Learn More
Bollé Safety Cobra ESP Safety Glasses
Product Code. 15538061
Product Code. 15538061
Brand: Bollé Safety COBESP
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Offers 180°; vision and perfect optical quality. Bollé™ COBRA Wrap-around Safety Glasses are the essential all rounder with panoramic visual field.
  • One product = two uses
  • Upper protection
  • Panoramic vision
  • Non-slip bridge
  • Comfortable, non-slip straight temples
  • Temple or strap version with interchangeable foam
  • Pivoting temples
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Frame: EN 166 BT CE; Lens marking: 5-1.4 1 BT CE; Chemical risk - Liquid droplets - 3 (Goggle, adapted spectacles with strap or face shield mandatory); Mechanical risk - Low energy impact - F; Mechanical risk- High speed particles at extreme temperatures
Lens Coating Anti-fog, Scratch-resistant
Description COBRA wrap around safety glasses
For Use With (Equipment) Black
Includes B-ESP filters; Scratch-resistant, anti-fog (ASAF)
No. per Pack 1 Ea.
Weight (Metric) 25 g
Lens Material Panoramic vision
Frame Material Wraparound, non-slip bridge