missing translation for 'onlineSavingsMsg'
Learn More

Cocamidopropyl Betaine (~30% aqueous solution), TRC

Product Code. 17991415
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17991415 10 g 10g
17990805 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17991415 Supplier Toronto Research Chemicals Supplier No. TRCC63351310G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86438-79-1
Molecular Formula C19H38N2O3
Molecular Weight (g/mol) 342.52
Synonym Cocamidopropyl Betaine
IUPAC Name 2-((3-dodecanamidopropyl)dimethylammonio)acetate
SMILES CCCCCCCCCCCC(NCCC[N+](C)(C)CC([O-])=O)=O

Specifications

Concentration ~30%
Quantity 10 g
InChI Formula InChI=1S/C19H38N2O3/c1-4-5-6-7-8-9-10-11-12-14-18(22)20-15-13-16-21(2,3)17-19(23)24/h4-17H2,1-3H3,(H-,20,22,23,24)
Formula Weight 342.29
Concentration or Composition (by Analyte or Components) [3-(Dodecanoylamino)propyl](dimethyl)ammonioacetate ~30%
Chemical Name or Material [3-(Dodecanoylamino)propyl](dimethyl)ammonioacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.