missing translation for 'onlineSavingsMsg'
Learn More

(-)-Coclaurine Hydrochloride, TRC

Product Code. 30134647
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134647 10 mg 10mg
30009616 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134647 Supplier Toronto Research Chemicals Supplier No. TRCC63355010MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3422-42-2
Molecular Formula C17H20ClNO3
Molecular Weight (g/mol) 321.8
Synonym 1,2,3,4-Tetrahydro-1-[(4-hydroxyphenyl)methyl]-6-methoxy-7-isoquinolinol
IUPAC Name (1S)-1-[(4-hydroxyphenyl)methyl]-6-methoxy-1,2,3,4-tetrahydroisoquinolin-7-ol;hydrochloride
SMILES COC1=C(C=C2C(=C1)CCN[C@H]2CC3=CC=C(C=C3)O)O.Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H19NO3.ClH/c1-21-17-9-12-6-7-18-15(14(12)10-16(17)20)8-11-2-4-13(19)5-3-11;/h2-5,9-10,15,18-20H,6-8H2,1H3;1H/t15-;/m0./s1
Formula Weight 321.1132
Chemical Name or Material (-)-Coclaurine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.