missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ COD Cuvette Tubes

Product Code. 10714087
Change view
Click to view available options
Range:
15 to 150mg/L
5 to 60mg/L
Unit Size:
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
10714087 15 to 150mg/L Pack of 25
10415815 5 to 60mg/L Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10714087 Supplier Hach Lange™ Supplier No. LCK314
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Standard pre-dosed, ready to use, pack of 25 COD Cuvette tubes features convenient and error-free dosing of reagents and provides precise and reliable measurements of values.

    • COD chemical oxygen demand cuvette tube cell vial test 15 - 150mg/L O2 (pack of 25)
  • For use with DR3900, DR6000, DR1900, DR2800, DR3800, and DR5000 instruments
  • Cuvette test, (tube test / cell test / vial test)
  • Photometric, chemical oxygen demand (COD)
  • 15 - 150mg/L O2 measurement range
  • Dichromate method according to ISO 6060-1989 standard and DIN 38409-H41-H44
  • Standard pre-dosed, ready to use, pack of 25 
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 6060-1989, DIN 38409-H41-H44
Quantity 25 Pack
Range 15 to 150mg/L
Type COD Cuvette Tube
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • CSB/COD/DCO
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric acid, Mercury(II) sulfate, Silver sulfate, potassium dichromate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.