missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ COD Reagent Test Tubes

Product Code. 11918437
Change view
Click to view available options
Range:
0 to 2000mg/L
Description:
Mercury Free
With Mercury
Unit Size:
25 tests
Pack of 24
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Description Unit Size
11918437 0 to 2000mg/L Mercury Free 25 tests
11946473 0 to 2000mg/L With Mercury Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11918437 Supplier Palintest™ Supplier No. PL454
  • ONLINE EXCLUSIVE PRICE
    108.63 €
    70.82 € / 25 tests
    Save 37.81 €
    34% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

These COD reagent test tubes, with a range of 0 to 2000mg/L, are ideal for use in most laboratory and scientific environments where measuring COD.

  • Ready to use reagent test tubes
  • COD reagent test tubes
  • Quantity: 1
  • Number of Tests: 25
  • Range: 0 to 2000mg/L
  • Type: COD reagent test tubes

Specifications

Description Mercury Free
No. of Tests 25
Quantity 1 Each
Range 0 to 2000mg/L
Type COD Reagent Test Tube
compliance-icons compliance-icons compliance-icons
Product Identifier
  • COD 2000 Solution
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P363-Wash contaminated clothing before reuse.
  • P391-Collect spillage.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: sulphuric acid, silver sulphate, potassium dichromate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.