missing translation for 'onlineSavingsMsg'
Learn More

Coelenterazine, native, Aequorea sp., Thermo Scientific Chemicals

Product Code. 15428339
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428339 1 mg 1mg
15438339 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428339 Supplier Thermo Scientific Alfa Aesar Supplier No. J66823.M^
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A sensitive and specific chemiluminescence probe of the superoxide anion

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55779-48-1
Molecular Formula C26H21N3O3
Molecular Weight (g/mol) 423.472
MDL Number MFCD00467176
InChI Key YHIPILPTUVMWQT-UHFFFAOYSA-N
Synonym coelenterazine, oplophorus luciferin, unii-3o1cb88rrd, clzn, coelenterate luciferin, coelenterazine, native, 3o1cb88rrd, 8-benzyl-2-4-hydroxybenzyl-6-4-hydroxyphenyl imidazo 1,2-a pyrazin-3 7h-one, 2-p-hydroxybenzyl-6-p-hydroxyphenyl-8-benzylimidazo 1,2-a pyrazin-3-7h-one, 3,2-dihydro-2-p-hydroxybenzyl-6-p-hydroxyphenyl-8-benzylimidazolo 1,2-a pyrazin-3-one
PubChem CID 2830
ChEBI CHEBI:2311
IUPAC Name 8-benzyl-6-(4-hydroxyphenyl)-2-[(4-hydroxyphenyl)methyl]-7H-imidazo[1,2-a]pyrazin-3-one
SMILES C1=CC=C(C=C1)CC2=C3N=C(C(=O)N3C=C(N2)C4=CC=C(C=C4)O)CC5=CC=C(C=C5)O

Specifications

Packaging Plastic bottle
Quantity 1 mg
Sensitivity Light and moisture sensitive
Solubility Information Soluble in methanol or ethanol
Formula Weight 423.46
Physical Form Solid
Chemical Name or Material Coelenterazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.