Learn More
Abcam Coenzyme A Assay Kit (Fluorometric - Green)
Product Code. 32118271 Shop All Abcam Products
Change view
Click to view available options
200 Tests
Product Code.
32118271 200 Tests
1 options
Product Code. 32118271 Supplier Abcam Supplier No. ab138889200Test
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Measure coenzyme A in 1h in Cell/Tissue Extracts with Coenzyme A Assay Kit AB138889. Sensitivity = 40 nM. Cited in 8 publications. Use with Microplate reader.
Measure coenzyme A in 1h in Cell/Tissue Extracts with Coenzyme A Assay Kit AB138889. Sensitivity = 40 nM. Cited in 8 publications. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 200 Tests