missing translation for 'onlineSavingsMsg'
Learn More

Coenzyme A Trilithium Salt Dihydrate ≥96%, White Crystalline Powder, MP Biomedicals™

Product Code. 11497540 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
250 mg
500 mg
Unit Size:
100mg
10mg
250mg
25mg
500mg
50mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497540 50 mg 50mg
11477540 10 mg 10mg
11487540 25 mg 25mg
11407550 100 mg 100mg
11417550 250 mg 250mg
11427550 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11497540 missing translation for 'mfr' MP Biomedicals missing translation for 'supplierNo' 0210049350
  • ONLINE EXCLUSIVE PRICE
    256.00 €
    242.55 € / 50mg
    Save 13.45 €
    5% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 18439-24-2
Molecular Formula C21H33Li3N7O16P3S
Molecular Weight (g/mol) 785.329
MDL Number MFCD00075848
InChI Key QSCBPHBAFBVXRK-UHFFFAOYSA-K
Synonym coenzyme a, trilithium salt
PubChem CID 53442190
IUPAC Name trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate
SMILES [Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O

Specifications

Color White
Assay Percent Range ≥96%
Quantity 50 mg
Formula Weight 821.4
Physical Form Crystalline Powder
Chemical Name or Material Coenzyme A Trilithium Salt Dihydrate >96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.