missing translation for 'onlineSavingsMsg'
Learn More

Coenzyme A Trilithium Salt Dihydrate, MP Biomedicals™

Product Code. 11437550 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11437550 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11437550 Supplier MP Biomedicals Supplier No. 0210049391
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Facilitates removal of lipid peroxides and promotes repair of plasma membranes. Coenzyme A Trilithium Salt Dihydrate, MP Biomedicals is an essential cofactor in enzymatic acetyl transfer reactions. It increases mobilization of fatty acids and activates ph

The principal biologically active forms of pantothenic acid are coenzyme A (CoA) and acyl carrier protein (ACP). In CoA, the business center of the molecule is the pantothenic acid metabolite 4'-phosphopantetheine.

  • Coenzyme A is comprised of 4'-phosphopantetheine linked by an anhydride bond to the nucloetide adenosine 5'-monophosphate
  • 4'-Phosphopantetheine itself is comprised of pantothenic acid linked at one end, via an amide bond, to beta-mercaptoethylamine, derived from L-cysteine, and at the other end to a phosphate group
  • The sulfhydryl group of 4'-phosphopantetheine, which is the business end of the coenzyme, forms thioesters with acyl groups producing acyl-CoA derivatives, including acetyl-CoA

Specifications

CAS 18439-24-2
Molecular Formula C21H33Li3N7O16P3S
Quantity 1 g
Synonym coenzyme a, trilithium salt
Solubility Information Soluble in water (50mg/mL, clear, colorless to faint yellow solution)
InChI Key QSCBPHBAFBVXRK-UHFFFAOYSA-K
SMILES [Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O
IUPAC Name trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate
Molecular Weight (g/mol) 785.329
PubChem CID 53442190
Formula Weight 821.4
Percent Purity ≥96%
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.