missing translation for 'onlineSavingsMsg'
Learn More

Virkon™ Cold Liquid Sterilization

Product Code. 15196603
Change view
Click to view available options
Quantity:
12 Pk.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15196603 12 Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15196603 Supplier Virkon™ Supplier No. 332003
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Top level disinfectant of invasive medical devices and instruments

  • Invasive medical equipment sterilant
  • Ideal for fibre optic instruments and probes
  • The best top level disinfectant of invasive medical devices and instruments such as: fibre optical instruments; surgical probes, etc.

Specifications

Product Type Cold Liquid Sterilization
Color-coded No
Form Powder
Scent Regular Scent
Sterility Sterile
Quantity 12 Pk.
Weight (Metric) 162 g
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Perasafe Sterilant / Disinfectant 16.2g Sachet
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Self-reactive substance or mixture Type E & F
Hazard Statement
  • H242-Heating may cause a fire.
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P235-Keep cool.
  • P264-Wash skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403-Store in a well-ventilated place.
  • P420-Store away from other materials.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: disodium carbonate,compound with hydrogen peroxide, citricacid, sodium carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.