Learn More
KGW Isotherm™ Cold Trap
Product Code. 10202022
Change view
Click to view available options
Model:
KF 29-GL
KF 29-GL-A
KFL 29-GL
Includes:
12C Dewar
12C Dewar, Spout
18C Dewar
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Includes Unit Size
10202022 KF 29-GL 12C Dewar Each
10789444 KFL 29-GL 18C Dewar Each
10264892 KF 29-GL-A 12C Dewar, Spout Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10202022 Supplier KGW Isotherm™ Supplier No. 1740
  • ONLINE EXCLUSIVE PRICE
    583.00 €
    503.32 € / Pack of 1
    Save 79.68 €
    13% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Capacity (Metric) 150 mL (Condensate), 1 L (Coolant)
Includes 12C Dewar
Quantity 1 Pack
Type Cold Trap (Short)
Model KF 29-GL
Connection Type Glass Thread GL18 with Olive