Learn More
KGW Isotherm™ Cold Trap
Product Code. 10789444
Change view
Click to view available options
Model:
KF 29-GL
KF 29-GL-A
KFL 29-GL
Includes:
12C Dewar
12C Dewar, Spout
18C Dewar
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Includes unitSize
10789444 KFL 29-GL 18C Dewar Each
10202022 KF 29-GL 12C Dewar Each
10264892 KF 29-GL-A 12C Dewar, Spout Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10789444 Supplier KGW Isotherm™ Supplier No. 17362
Only null left
Add to basket
Add to basket
Specifications
Capacity (Metric) 250 mL (Condensate), 2 L (Coolant)
Includes 18C Dewar
Quantity 1 Pack
Type Cold Trap (Long)
Model KFL 29-GL
Connection Type Glass Thread GL18 with Olive