Learn More
VACUUBRAND™ Cold Trap
Product Code. 10548381
Change view
Click to view available options
Type:
GKF 1000i Cold Trap
SKF H 25 Cold Trap
SKF H 40 Cold Trap
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10548381 GKF 1000i Cold Trap Each
10428181 SKF H 25 Cold Trap Each
10524902 SKF H 40 Cold Trap Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10548381 Supplier VACUUBRAND™ Supplier No. 20667056
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use this Cold Trap with very volatile solvents and reduce the back streaming of oil molecules into the vacuum system. Vacubrand™ Cold Traps provide important protection for your pump by separating condensates and aggressive media.
Cold trap SKF
  • Sturdy, easy to clean
  • Easy to disassemble
  • Two-wall design with good conductance
  • Long operating time per coolant filling
  • Easy condensate drainage without disassembling
Cold trap GKF
  • Highly reflective insulation jacket for extended coolant life
  • Vertical window: Direct observation of condensate and coolant levels
  • PTFE stopcock: Drain condensate without disassembly
  • Sheet metal shield for protection against external damage and implosion
Inline oil filter HF
  • Extended oil lifetime
  • Reduced service demands
  • Easy space-saving mounting
  • Simple filter change, service indicator
TRUSTED_SUSTAINABILITY
Specifications
Case Material Borosilicate Glass, PTFE, FKM
For Use With (Equipment) Vacuubrand pumps
Type GKF 1000i Cold Trap