Learn More
Buerkle™ Collection Cylinder for Volume Heads PharmaPicker
Product Code. 31188900 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
0.1 to 1.2 mL
1.25 to 2.5 mL
Product Code. 31188900
Brand: Buerkle™ 53461001
The collection chamber is the sample intake cylinder of the PharmaPicker system. The corresponding volume tip is inserted into the front of this stainless steel chamber. When inserted into the powder, the tip fills up, as it is withdrawn, the sample volume remains in the collection chamber.
The collection chamber is available in two sizes, suited to the respective volume range of the tips. It features a smooth surface that can be cleaned without residue and sterilised. The collection chamber ensures that the collected powder sample is securely stored until it is analysed and enables rapid volume changes by simply exchanging the tips.
  • Stainless steel collection cylinder for PharmaPicker - container for the collected sample
TRUSTED_SUSTAINABILITY
Specifications
Product Type Sampler Insert
Material Stainless Steel (V4A)
Autoclavable Autoclavable
Capacity (Metric) 0.1 to 1.2 mL