missing translation for 'onlineSavingsMsg'
Learn More

SCAT Collectors for HPLC

Product Code. 10590251 Shop All SCAT Europe Products
missing translation for 'orderingAttributeHoverText'
Product Type:
2-in-1 Connector
3-in-1 Connector
3-in-1 Sideways Connector
3-in-1 Vertical Connector
8-in-1 Connector
missing translation for 'unitSize'
Each
Diameter (Metric) Inner:
5.0 to 11.5 mm
Diameter (Metric) Outer:
3.2 mm
This item is not returnable. View return policy

Product Code. 10590251

missing translation for 'mfr': SCAT 199010

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Collect several tubes into a single connector - no problem with the right collector! The tube connector is the most flexible connector of the SCAT system.

For a reliable and clean connection of hoses and capillaries for collecting liquids. SCAT collectors are available in versions with up to 8 connection options.The connections are either on the side or vertically, so that the tubes can be connected easily. Tubes with different diameters from 2.3 to 11.5 mm can be connected. The connectors are made of chemical-resistant PTFE, PFA or PP material.

Specifications

Autoclavable Autoclavable
Material PTFE
For Use With Hitachi-Chromaster HPLC System
Includes Fittings for capillaries of 2.3/3.2 mm, 3-in-1 Connector
Color White
Product Type 3-in-1 Connector
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.