Learn More
Lenz Laborglasintrumente Columns Acc. to Hempel with Removable Glass Jacket
Product Code. 10498402
Click to view available options
Length (Metric):
200 mm
300 mm
500 mm
800 mm
Unit Size:
Each
Product Code. 10498402
Brand: Lenz Laborglasintrumente 05117301
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Columns acc. to Hempel
  • With removable glass jacket
  • For packings like Raschig rings or glass beads
  • With NS socket and NS cone
  • With removable glass jacket
  • Made of DURAN™ tubing
Specifications
Includes Removable glass jacket
Length (Metric) 300 mm
Standard Taper Joint NS 14/23