missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Comacier VHP Plus Heavy Duty Leather Gloves

Product Code. 15555746
Click to view available options
Size:
8
9
10
11
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 15555746

Brand: Ansell™ 97039080

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provides protection against cuts with flexibility and comfort. Ansell Comacier VHP Plus Heavy Duty Leather Gloves are made from high performance fibres using Techcor™ Knitting Technology.

  • Mechanical protection heavy duty gloves
  • Medium weight 10 gauge seamless knitted glove
  • Knit wrist ensures good fit
  • Unique traceability system on textile tag
  • Palm, fingers and artery guard made of leather for optimal protection
  • Provides the highest levels of mechanical resistance
  • Good flexibility and comfort
  • Superior grip when used with oils and greases due to leather patch

Applications:

  • Handling heavy and oily metal pieces
  • Handling sharp or irregular-edged metal parts
  • Press and stamping
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 388:2003
Color Gray
Exterior Finish Leather Palm
Latex Free No
Material Leather
Product Type Mechanical Protection Gloves
Size 8
Glove Cuff Style Knit
Liner Material Techcor™
Packaging Type 12 pairs in bag, 10 bags in carton
Palm Type Leather
Series VHP Plus
Length (Metric) 240 to 260 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.