missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Combitips advanced™ Rack
GREENER_CHOICE

Product Code. 12654607 Shop All Eppendorf Products
Change view
Click to view available options
Holds:
Combitips (0.1 to 10 mL)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
12654607 Combitips (0.1 to 10 mL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12654607 Supplier Eppendorf™ Supplier No. 0030089758

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use this rack with Combitips advanced™ pipette tips. Eppendorf™ Combitips advanced™ Rack holds 8 combitips advanced pipette tips.

Use this rack with Combitips advanced™ pipette tips. Eppendorf™ Combitips advanced™ Rack holds 8 combitips advanced pipette tips.

For 0.1 to 10 mL tips

TRUSTED_SUSTAINABILITY

Specifications

Holds Combitips (0.1 to 10 mL)
Positions 8
For Use With (Application) Single-handed dispensing
For Use With (Equipment) 8 Combitips advanced™ (0.1 to 10mL)
Quantity 100 Pack
Product Type Combitip Rack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.