missing translation for 'onlineSavingsMsg'
Learn More

Combretastatin A4 Phosphate Disodium Salt, TRC

Product Code. 30364472
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364472 25 mg 25mg
30365428 10 mg 10mg
30365684 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364472 Supplier Toronto Research Chemicals Supplier No. TRCC64302525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 168555-66-6
Molecular Formula C18 H19 O8 P . 2 Na
Molecular Weight (g/mol) 440.29
IUPAC Name disodium;[2-methoxy-5-[(Z)-2-(3,4,5-trimethoxyphenyl)ethenyl]phenyl] phosphate
SMILES [Na+].[Na+].COc1ccc(\C=C/c2cc(OC)c(OC)c(OC)c2)cc1OP(=O)([O-])[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H21O8P.2Na/c1-22-14-8-7-12(9-15(14)26-27(19,20)21)5-6-13-10-16(23-2)18(25-4)17(11-13)24-3;;/h5-11H,1-4H3,(H2,19,20,21);;/q;2*+1/p-2/b6-5-;;
Formula Weight 440.0613
Chemical Name or Material Combretastatin A-4 Phosphate, Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.