missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for omniPAGE Mini Vertical Gel Electrophoresis Unit, 0.5mm Thick

Product Code. 11887603 Shop All Fisherbrand Products
Change view
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
12
16
20
5
8
9
Volume (Metric) Well:
10 μL
12.5 μL
16 μL
20 μL
25 μL
30 μL
50 μL
330 μL
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells Volume (Metric) Well unitSize
11887603 5 50 μL Each
11857583 10 20 μL Each
11837593 12 16 μL Each
11887593 16 12.5 μL Each
11837603 20 10 μL Each
11837613 8 30 μL Each
11887613 9 25 μL Each
11837583 1 Prep, 1 Marker 330 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11887603

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

For use in preparatory electrophoresis and high-throughput screening.

  • Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
  • 8-sample and 16-sample combs are Multichannel pipettor compatible

Specifications

Color Purple
For Use With (Application) Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations
For Use With (Equipment) OmniPAGE Mini Vertical Gel Electrophoresis Unit
Type Comb
No. of Wells 5
Quantity 1 Each
Thickness (Metric) 0.5 mm
Volume (Metric) Well 50 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.