missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for MultiSUB Maxi Horizontal Gel Electrophoresis Unit, 1.5mm Thick

Product Code. 11893443 Shop All Fisherbrand Products
Change view
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
16
2 Prep, 2 Marker
20
25
30
36
4 Prep, 2 Marker
40
50
Volume (Metric) Sample:
16 μL
17 μL
22 μL
26 μL
32 μL
41 μL
61 μL
108 μL
230 μL
473 μL
1013 μL
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells Volume (Metric) Sample unitSize
11893443 30 26 μL Each
11863423 10 108 μL Each
11893423 1 Prep, 1 Marker 1013 μL Each
11833433 16 61 μL Each
11883433 20 41 μL Each
11813443 2 Prep, 2 Marker 473 μL Each
11853443 25 32 μL Each
11833453 36 22 μL Each
11883453 40 17 μL Each
11813463 4 Prep, 2 Marker 230 μL Each
11853463 50 16 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 11893443
  • ONLINE EXCLUSIVE PRICE
    50.25 €
    35.93 € / Pack of 1
    Save 14.32 €
    28% Off
Estimated Shipment: 11-09-2026
Add to basket
Add to basket

For use in preparatory electrophoresis and high-throughput screening.

  • Recommended for high resolution gels and publication quality data
  • Maximise sample volume
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 20-sample and 40-sample combs are Multichannel pipettor compatible

Specifications

Color Red
For Use With (Equipment) MultiSUB Maxi Horizontal Gel Electrophoresis Unit
Type Comb
No. of Wells 30
Quantity 1 Each
Thickness (Metric) 1.5 mm
Volume (Metric) Sample 26 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.