missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 2mm Thick

Product Code. 11813403 Shop All Fisherbrand Products
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
12
14
16
18
2 Prep, 2 Marker
20
28
30
35
4 Prep, 2 Marker
Volume (Metric) Sample:
20 μL
23 μL
25 μL
43 μL
54 μL
59 μL
81 μL
90 μL
243 μL
450 μL
990 μL
Unit Size:
Each

Product Code. 11813403

Only null left
Add to basket
Add to basket

For use in preparatory electrophoresis and high-throughput screening.

  • Recommended for high resolution gels and publication quality data
  • Maximise sample volume
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10, 14, 16, 18, 28 and 30-sample combs are Multichannel pipettor compatible

Specifications

Color Blue
For Use With (Equipment) MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit
Type Comb
No. of Wells 28
Quantity 1 Each
Thickness (Metric) 2 mm
Volume (Metric) Sample 23 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.