Learn More
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 1mm Thick
Product Code. 11857593 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
12
16
20
5
8
9
Volume (Metric) Well:
20 μL
25 μL
35 μL
40 μL
50 μL
60 μL
100 μL
650 μL
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells Volume (Metric) Well Unit Size
11857593 12 35 μL Each
11877583 10 40 μL Each
11807593 1 Prep, 1 Marker 650 μL Each
11807603 16 25 μL Each
11857603 20 20 μL Each
11807613 5 100 μL Each
11857613 8 60 μL Each
11807623 9 50 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11857593
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use in preparatory electrophoresis and high-throughput screening.
  • Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
  • Supplied as standard
  • 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
For Use With (Application) Mini SDS PAGE, Native PAGE, Gradient, Second Dimension and Nucleic Acid Separations
For Use With (Equipment) OmniPAGE Mini Vertical Gel Electrophoresis Unit
Type Comb
No. of Wells 12
Quantity 1 Each
Thickness (Metric) 1 mm
Volume (Metric) Well 35 μL