Learn More
Thermo Scientific™ Combustible Gas Tap for Rear Wall
Product Code. 10686473 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Laboratory Enclosure
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
10686473 Laboratory Enclosure Each
1 options
Product Code. 10686473 Supplier Thermo Scientific™ Supplier No. 50068235
  • ONLINE EXCLUSIVE PRICE
    279.39 €
    252.00 € / Pack of 1
    Save 27.39 €
    9% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For Herasafe 2025, 2030i and Maxisafe 2030i Biological Safety Cabinet
For Herasafe 2025, 2030i and Maxisafe 2030i Biological Safety Cabinet
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Laboratory Enclosure
Product Type Add-A-Valve Kit for Main Chamber