missing translation for 'onlineSavingsMsg'
Learn More

Comfort Black Nitrile Examination Gloves

Product Code. 16652227
Click to view available options
Size:
Large
X-Large
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 16652227

Brand: New source - PPE PFNBR 244 L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Single use, black nitrile examination gloves offer excellent protection for general lab use. They are particularly suitable for industrial, mechanical and technical environments where staining may be an issue. Nitrile gloves are a good alternative for users with latex allergies.

  • Non-sterile
  • Disposable
  • Powder free
  • Textured palm for excellent grip
  • Ambidextrous
  • Distinctive black color hides stains
  • AQL 1.5

Specifications

Certifications/Compliance AQL 1.5, EN 374-1:2016, EN374-2:2014, EN 374-4:2013, EN374-5:2016, EN 455 part 2, EN 420:2003+A1, ASTM D6124
Description Shelf Life 3 Years
Color Black
Material Nitrile
Size Large
Thickness (Metric) 0.07 ±0.02 mm (Cuff)
Quantity 100 Gloves/Box
Yarn Weight 4.8 gm ± 0.2 gm
Length (Metric) 240 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.