missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Comfort Rack for Upright Freezers, 50 mm Box Height
Comfort Rack designed to ensure a more ergonomical operating process when working with samples.
160.00 € - 205.00 €
Specifications
Height (Metric) | 50 mm (Box) |
---|---|
For Use With (Equipment) | Upright Freezers |
Material | Stainless Steel |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
15210734
|
Tenak™
TE24220 |
Boxes (9) |
160.00 €
Each |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
15230744
|
Tenak™
TE24236 |
Boxes (24) |
186.00 €
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||||
15240754
|
Tenak™
TE24256 |
Boxes (30) |
205.00 €
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
15260734
|
Tenak™
TE24226 |
Boxes (18) |
179.00 €
Each |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
15270734
|
Tenak™
TE24228 |
Boxes (21) |
184.00 €
Each |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
15270744
|
Tenak™
TE24250 |
Boxes (15) |
177.00 €
Each |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
15280734
|
Tenak™
TE24230 |
Boxes (12) |
162.00 €
Each |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
Description
- In the comfort rack, the cryoboxes are placed in drawers
- Increases the flexibility and reduces the operating weight compared to the rack types
- Measure the inside height and depth of you freezer when choosing the right rack
- Racks can be used with all freezer models
- Constructed of stainless steel
Specifications
50 mm (Box) | |
Upright Freezers | |
Stainless Steel |