missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caframo™ Light Torque Overhead Stirrers 220V
Description
- Stand, clamp and stirring impeller are sold separately.
- Stirlight: Illuminate solutions and dimly lit workstations
- Digital display accurate to 1rpm
- Last speed used stored when stirrer is turned off
- Through shaft for easy impeller adjustment
- Overload protection
- Clockwise and counter clockwise rotation on Reversing Digital BDC2010U1
- Brushless DC motor provides maximum power and is maintenance free
- AutoStart and Timer features
- Safety approved: CE marked Applications include: chemical mixing in a fume hood or enclosed workstation, pharma, suspensions, medical labs, serum prep, blood product mixing and reactions.
Specifications
Specifications
| Speed | 40 to 2010 rpm |
| Capacity | 25 L |
| Display | Digital |
| Voltage | 220/230 V |
| Wattage | 70 w |
| Height (English) | 9.7 in. |
| Height (Metric) | 24.7 cm |
| Length (English) | 11 in. |
| Length (Metric) | 27.9 cm |
| Frequency | 50/60 Hz |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?