missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Compact Digital Dry Bath/Block Heater

Product Code. 15312225 Shop All Thermo Scientific Products
Change view
Click to view available options
Model:
Compact Dry Bath D
Compact Dry Bath S
Plug Type:
CN, EU, UK
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Plug Type Unit Size
15312225 Compact Dry Bath D CN, EU, UK Each
15302225 Compact Dry Bath S CN, EU, UK Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15312225 Supplier Thermo Scientific™ Supplier No. 88871004
  • ONLINE EXCLUSIVE PRICE
    550.00 €
    479.75 € / Pack of 1
    Save 70.25 €
    12% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Maximize bench space with our compact, lightweight dry bath with multiple combinations of block sizes for versatility.

Maximize valuable bench space with Thermo Scientific™ Compact Digital Dry Bath/ Block Heater—available with multiple combinations of block sizes for tube size versatility.

  • Built-in temperature-sensing probe assists with outstanding temperature accuracy and control
  • Designed for precise temperature control
  • Fast heating rate
  • Multiple combinations of different sizes of blocks allows for maximum tube size versatility during experimental procedure
  • 12V low voltage power input
  • Two sizes to meet your application volume demands
  • Convenient and easy-to-read digital display
  • Small footprint
  • Lightweight design
  • Single (S) model can hold a full block, 2 half blocks, 4 quarter blocks or 1 half and 2 quarter blocks
  • Double (D) model can hold 2 full blocks, 4 half blocks, 8 quarter blocks or a combination of full/half and quarter blocks.

Applications:

  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification

Required Accessory:

  • Unit must be ordered with block(s)

Warranty

1 Year, Replacement

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, cTUVus, RoHS
Controller Type Digital
Capacity (Metric) 0.2 mL
Lighting LED
Plug Type CN, EU, UK
Temperature Range (Metric) Ambient +2° to 100°C
Type Dry Block Heater
Heating Rate 20 min. average heating rate (30° to 100°C, ambient at 25°C)
Includes Dry bath, CN, EU,and UK style plugs, block lifter, plastic lid
Model Compact Dry Bath D
No. of Blocks 2 full blocks, 4 half blocks, 8 quarter blocks or a combination of full/half and quarter blocks
Temperature Accuracy ≤ ± 0.5°C
Temperature Uniformity ≤ ± 0.5°C
Warranty 1 Year, Replacement
Dimensions (L x W x H) 200 x 200 x 25 mm (7.9 x 7.9 x 1 in.)
Length (English) 7.9 in.
Length (Metric) 200 mm
Width (English) 7.9 in.
Width (Metric) 200 mm
Height (English) 1 in.
Height (Metric) 25 mm
Weight (English) 3.3 lb.
Weight (Metric) <1.5 kg
Electrical Requirements 12 V, DC, 7 A
Voltage 100/240 V
Show More Show Less

Warranty: 1-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.