missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Compact Digital Dry Bath/Block Heater
334.92 EUR valid until 2024-12-31
Use promo code "21906" to get your promotional price.
300.00 EUR valid until 2024-03-30
Use promo code "21648" to get your promotional price.
334.92 EUR valid until 2024-04-30
Use promo code "21656" to get your promotional price.
334.92 EUR valid until 2024-03-30
Use promo code "21741" to get your promotional price.
Use promo code "21906" to get your promotional price.
300.00 EUR valid until 2024-03-30
Use promo code "21648" to get your promotional price.
334.92 EUR valid until 2024-04-30
Use promo code "21656" to get your promotional price.
334.92 EUR valid until 2024-03-30
Use promo code "21741" to get your promotional price.
Click to view available options
Model:
Compact Dry Bath D
Compact Dry Bath S
Heating Rate:
20 min. average heating rate (30° to 100°C, ambient at 25°C)
Plug Type:
CN, EU, UK
Description
Maximize valuable bench space with Thermo Scientific™ Compact Digital Dry Bath/ Block Heater—available with multiple combinations of block sizes for tube size versatility.
- Built-in temperature-sensing probe assists with outstanding temperature accuracy and control
- Designed for precise temperature control
- Fast heating rate
- Multiple combinations of different sizes of blocks allows for maximum tube size versatility during experimental procedure
- 12V low voltage power input
- Two sizes to meet your application volume demands
- Convenient and easy-to-read digital display
- Small footprint
- Lightweight design
- Single (S) model can hold a full block, 2 half blocks, 4 quarter blocks or 1 half and 2 quarter blocks
- Double (D) model can hold 2 full blocks, 4 half blocks, 8 quarter blocks or a combination of full/half and quarter blocks.
Applications:
- Sample preparation
- Enzyme preservation
- Enzyme-substrate reactions
- DNA amplification
- Unit must be ordered with block(s)
Warranty
1 Year, Replacement
Specifications
Certifications/Compliance | CE, cTUVus, RoHS |
Controller Type | Digital |
Capacity (Metric) | 0.2 mL |
Lighting | LED |
Plug Type | CN, EU, UK |
Temperature Range (Metric) | Ambient +2° to 100°C |
Type | Dry Bath/Block |
Heating Rate | 20 min. average heating rate (30° to 100°C, ambient at 25°C) |
Includes | Dry bath, CN, EU,and UK style plugs, block lifter, plastic lid |
Model | Compact Dry Bath S |
Show More |
Warranty: 1-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'