missing translation for 'onlineSavingsMsg'
Learn More

Compbead anti-Rat and anti-Hamster Ig κ/Negative Control Compensation Particles Set, BD

Produktkod. 15507465
Change view
Klicka för att se tillgängliga alternativ
Quantity:
6 mL Each
Förpackningsstorlek:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Förpackningsstorlek
15507465 6 mL Each 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15507465 Leverantör BD Biosciences Leverantörsnummer 552845
 Restricted Item
Estimated Shipment: 08-10-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

The BD CompBeads Compensation Particles Anti-Rat/Hamster Ig, kappa Set are polystyrene microparticles which are used to optimize fluorescence compensation settings for multicolor flow cytometric analyses. The set provides two populations of microparticles, the BD CompBeads Anti-Rat/Hamster Ig, kappa particles, which bind any rat or hamster kappa light chain-bearing immunoglobulin, and the BD CompBeads Negative Control (FBS) Particles, which has no binding capacity. When mixed together with a fluorochrome-conjugated rat or hamster antibody, the BD CompBeads provide distinct positive and negative (background fluorescence) stained populations which can be used to set compensation levels manually or using instrument set-up software. Since the compensation adjustments are made using the same fluorochrome-labeled antibody to be used in the experiment, this method allows the investigator to more accurately establish compensation corrections for spectral overlap for any combination of fluorochrome-labeled antibodies (without having to use valuable tissue samples or hard-dyed beads with potentially mismatched fluorescence spectra). Use of the BD CompBeads is highly recommended for use in all experiments using tandem dye (i.e., PE-Cy7, APC-Cy7, etc.) conjugates, which may have distinct spectral characteristics for each conjugate.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Ig, κ
Conjugate Unconjugated
Purification Method Unpurified
Quantity 6 mL Each
Primary or Secondary Primary
Target Species Hamster, Rat
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.