missing translation for 'onlineSavingsMsg'
Learn More

Compbead anti-Rat Ig, κ/Negative Control (FBS) Compensation Particles Set, Clone: G16-510E3, BD

Product Code. 15896868
Change view
Click to view available options
Quantity:
6 mL Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896868 6 mL Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15896868 Supplier BD Biosciences Supplier No. 552844
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The BD CompBeads Compensation Particles Anti-Rat Ig, kappa Set contains polystyrene microparticles, which are used to optimizefluorescence compensation settings for multicolor flow cytometric analyses. The set provides two populations of microparticles, the Anti-Rat Ig, kappa particles, which bind any rat kappa light chain-bearing immunoglobulin, and the Negative Control particles that have no binding capacity. When mixed together with a fluorochrome-conjugated rat antibody, the BD CompBeads Compensation Particles provide distinct positive and negative (background fluorescence) stained populations which can be used to set compensation levels manually or automatically using instrument set-up software. Since the compensation adjustments are made using the same fluorochrome-labeled antibody to be used in the experiment, this method allows the investigator to more accurately establish compensation corrections for spectral overlap for any combination of fluorochrome-labeled antibodies without having to use valuable tissue samples or hard-dyed beads with potentially mismatched fluorescence spectra. Use of the BD CompBeads Compensation Particles is highly recommended for use in all experiments using tandem dye (i.e., PE-Cy7, APC-Cy7, etc.) conjugates, which may have distinct spectral characteristics for each conjugate.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat Ig, κ
Classification Monoclonal
Conjugate Unconjugated
Purification Method Unpurified
Quantity 6 mL Each
Primary or Secondary Primary
Target Species Rat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.