missing translation for 'onlineSavingsMsg'
Learn More

Abcam Compensation Beads Kit (3.0-3.4 μm) - separate vials

Product Code. 32115933 Shop All Abcam Products
Change view
Click to view available options
Quantity:
2 x 5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32115933 2 x 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32115933 Supplier Abcam Supplier No. ab3227002x5mL
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Compensation Beads Kit (3.0-3.4 μm) - separate vials (ab322700) is part of the sample prep & detection kits range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.

Compensation Beads Kit (3.0-3.4 μm) - separate vials (ab322700) is part of the sample prep & detection kits range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.

The product is subject to the following: Abcam Restricted Use Statement

Tekniske data

Quantity 2 x 5 mL
Regulatory Status RUO
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.