Specifications
| For Use With (Equipment) | XK 50 Columns |
| Material | Acetal plastic (POM); Ethylene propylene(EPDM); Nitrile rubber (NBR) |
| Type | Complete End Piece |
| Includes | End piece, O-ring (7.27 x 2.62 mm), Sealing ring with built-in stainless steel spiral spring |