Learn More
Thermo Scientific™ Complete Stainless Steel Rollout Drawer Set
Product Code. 15206615 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
11.5 cu. ft. Refrigerators
45.8 cu. ft. (1297L) Refrigerators and Freezers
No. of Drawers:
5
12
Unit Size:
Each
Pack of 12
Product Code. 15206615
Brand: Thermo Scientific™ 7445
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A convenient alternative to open-wire shelving
Choose stainless-steel Thermo Scientific™ Rollout Drawers as a convenient alternative to open-wire shelving. This is an option for Thermo Scientific™ high-performance refrigerators.
  • Stainless-steel drawers
  • Adjustable with full extension
Ordering Information:
  • Factory installed; must be specified when ordering the refrigerator
TRUSTED_SUSTAINABILITY
Specifications
Type Complete SS Drawer Set
Capacity (English) 11.5 cu. ft.
Capacity (Metric) 326 L
Drawers Stainless Steel (Drawer)
No. of Drawers 5
For Use With (Equipment) 11.5 cu. ft. Refrigerators