missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Compound E

Product Code. 18776179
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18776179 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18776179 Supplier R&D Systems™ Supplier No. 6476/1
  • ONLINE EXCLUSIVE PRICE
    482.90 €
    431.55 € / 1mg
    Save 51.35 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. γ-secretase inhibitor; induces neuronal differentiation

Compound E is a γ-secretase and notch patchway inhibitor. Compound E enhances growth inhibition, differentiation and migration of neuroblastoma cells when used in a combination with DAPT (Cat. No. 2634) and 13-cis RA. In combination with hLIF, CHIR 99021 (Cat. No. 4423) and SB 431542 (Cat. No. 1614), Compound E accelerates the induction of a homogeneous, self-renewing primitive neuroepithelium population from hESCs. Compound E is active in vitro and in vivo.

For more information about how Compound E may be used, see our protocol:

Generation of β cells from hPSCs
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(1S)-2-[[(3S)-2,3-Dihydro-1-methyl-2-oxo-5-phenyl-1H-1,4-benzodiazepin-3-yl]amino]-1-methyl-2-oxoethyl]-3,5-difluorobenzeneacetamide
CAS 209986-17-4
Quantity 1 mg
Product Line Tocris
Molecular Formula C27H24F2N4O3
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.