missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Con A Sepharose™ 4B Medium

Product Code. 11534905 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
Unit Size:
100mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11534905 5 mL 5mL
11524905 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11534905 Supplier Cytiva Supplier No. 17044003
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An affinity chromatography media for capture of glycosylated biomolecules including glycoproteins and glycolipids

Highly suited for isolation of cell surface glycoproteins from detergent-solubilized membranes.

  • Con A coupled to Sepharose, in a 4% agarose matrix via cyanogen bromide activation
  • Binds molecules that contain α-D-mannose, α-D-glucose, and sterically related residues with available C-3, C-4, or C-5 hydroxyl groups
  • Group-specific adsorbent for molecules containing sugars
  • Concentration of immobilized Con A, approx. 13mg/mL medium; binding capacity for porcine thyroglobulin, approx. 30mg/mL medium
  • Stable in the pH range 4 to 9, and in the presence of all commonly used buffers
  • Average particle size: 90μm
  • Delivered in 0.1M acetate buffer, pH 6.0, containing 1M NaCl, 1mM CaCl2, 1mM MgCl2, 1mM MnCl2, and 20% ethanol

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity 30mg/mL
Ligand Concentration 13mg Con A/mL Drained Medium
Matrix 4% Agarose
Particle Size 90 μm
pH 4 to 9
Quantity 5 mL
Content And Storage 2°C to 8°C
For Use With (Application) For separation and purification of Glycoproteins, Polysaccharides and Glycolipids, isolates cell surface Glycoproteins from detergent-solubilized membranes
Ligand Concanavalin A
compliance-icons
Product Identifier
  • Con A Sepharose 4B, 5 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.