Learn More
Pacific Handy Cutter Concealed Blade All Purpose Safety Cutter
Product Code. 17718977
Change view
Click to view available options
Color:
Yellow
Unit Size:
Pack of 50
Product Code. Color Unit Size
17718977 Yellow Pack of 50
1 options
Product Code. 17718977 Supplier Pacific Handy Cutter Supplier No. EBC1
  • ONLINE EXCLUSIVE PRICE
    301.40 €
    259.81 € / Pack of 50
    Save 41.59 €
    13% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The EBC1 is a particularly robust safety knife with a concealed blade. Therefore, it is particularly safe. It is perfect for cutting up to double-wall corrugated cardboard, strapping and many other materials.
  • A disposable product in hi-vis yellow. With practical tape splitter to minimise blade use and product damage.
  • Plus an eyelet for fastening a lanyard or hanging.
  • Do you need to cut to a greater distance? Then you will like our adapter DFCCON. This turns your EBC1 into a telescopic cutting tool.
Specifikationer
Product Type Knife
Disposable Single-use
Sterility Non-sterile
Color Yellow
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.