missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Gram’s Iodine Stain, Concentrated
Description
Pro-Lab Diagnostics™ stains are manufactured in purpose-designed laboratories to the highest standards using the finest raw materials. Strict quality control and filtering techniques are employed prior to release to ensure that all stains offer excellent performance characteristics.
- 10X concentrated – 100mL is diluted to 1L of ready-to-use stain
- Gram’s Iodine is also known as diluted Lugol’s Iodine
- Apply after staining with crystal violet, but before ethanol
- Stains the peptidoglycan of gram positive organisms
Specifications
Specifications
| Solution Type | Concentrated Reagent |
| Type | Gram Iodine |
| Quantity | 100mL |
| Certifications/Compliance | (EC) No 1907/2006, Annex II, (EU) No 453/2010, CE-mark |
| Format | Liquid |
| For Use With (Application) | Gram’s Staining Method for the Initial Differentiation of Gram Positive and Gram Negative Bacteria |
| Color | Dark brown |
| Composition | Ethanol, Potassium Iodide, Iodine, Methanol |
| Storage Requirements | Keep at temperature not exceeding 20°C |
| Stain Type | Gram Stain |
| Show More |
Safety and Handling
Product Identifier
- Grams Iodine Concentrate
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501-Dispose of contents/container to .
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- MIXTURE LIST-Contain: ethanol, potassium iodide, iodine, methanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?