missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MuA Transposase (concentrated)

Product Code. 10515559 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
22 μg
Unit Size:
20µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10515559 22 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10515559 Supplier Thermo Scientific™ Supplier No. F750C
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform functional analysis of proteins with these transposon-based tools.

transposon products are based on the transposition machinery of the bacteriophage Mu. During the lytic phase of the phage's life cycle the machinery replicates its genome by transposing repeatedly inside the host genome. The Mu transposition reaction has been modified into an in vitro reaction catalyzed by a single enzyme - MuA Transposase. In this system, one in vitro reaction is capable of generating more than a million transposon insertion clones.

Highlights

The Mutation Generation System (MGS Kit) and Stop Generation System (STOP Kit) were developed for functional analysis of proteins. These new transposon tools enable the creation of saturated libraries of mutated proteins in a single reaction with less hands-on time than any other method. The location of the transposon insertion in each mutated clone can be mapped by either PCR or sequencing. With MGS and STOP kits, thousands of mutated clones are ready for expression studies in just 2 to 3 days.

The MGS Kit contains the complete set of reagents for transposon-based linker scanning mutagenesis of any target protein. The MGS Entranceposons are designed for making subtle changes in the structure of a target protein by inserting 15 bp in-frame linkers throughout the corresponding target gene. This in-frame insertion allows for conservation of downstream sequences.

The STOPKit Entranceposons contain translational stop codons in all three reading frames within the terminal portion of the transposon sequence. The proprietary modification of the Stop Generation System makes it possible to generate a saturated C-terminal deletion library from virtually any target protein with a maximum addition of three amino acids.

Highlights

  • Efficient - Create saturated insertion libraries for sequencing and protein analysis in a single reaction
  • Fast - Decrease hands-on time compared to conventional methods
  • Random - Eliminate target site preference or insertion hot-spot
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.1 μg/μL
Form Solution
Format Tube
Reaction Speed Fast
Product Type Transposon Products
Quantity 22 μg
Starting Material DNA
Volume 20 μL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.