Learn More
Eppendorf™ Concentrator Plus Rotors
Designed with the customer in mind to deliver an ergonomic, intuitive and durable solution that fits comfortably right on your lab bench
457.00 € - 1230.00 €
Product Code | Brand | Capacity | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Description | Price | Quantity & Availability | ||||
10737274
|
Eppendorf™
5490037006 |
18 x 10 mL | Rotor F-40-18-19 |
580.00 €
Each |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10337133
|
Eppendorf™
5490036000 |
24 x 6 mL | Rotor F-45-24-12 |
457.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10125802
|
Eppendorf™
5490045009 |
2 x 96 x 0.2 mL | Rotor A-2-VC |
1230.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10609503
|
Eppendorf™
5490043006 |
16 x 6.5/10 mL | Rotor F-45-16-20 |
580.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10498983
|
Eppendorf™
5490040007 |
36 x 1.5 mL | Rotor F-40-36-12 |
580.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11960517
|
Eppendorf™
5490047001 |
6 x 15/50 mL | Rotor F-35-6-30 |
580.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10554244
|
Eppendorf™
5490038002 |
8 x 15 mL | Rotor F-45-8-17 |
580.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10327423
|
Eppendorf™
5490042000 |
8 x 16 mL | Rotor F-50-8-18 |
580.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10522844
|
Eppendorf™
5490041003 |
8 x 15 mL | Rotor F-50-8-16 |
580.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10391584
|
Eppendorf™
5490039009 |
8 x 25 mL | Rotor F-35-8-24 |
580.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Advanced heating technology provides best treatment for your sample which assures quick, efficient and gentle vacuum concentration of DNA/RNA, nucleotides, proteins and other liquid or wet samples. Our new coated lid provides superior chemical resistance against aggressive acids and organic solvents (e.g. TFA, DMSO).
- The Concentrator plus is available with three configurations to meet the customers' lab specific needs:
- Basic Concentrator for connecting to an existing vacuum pump
- Complete system with an integrated solvent-resistant, maintenance and oil-free PTFE pump, and condensation trap
- Complete system with vacuum port adds extra capability. It allows for using the integrated vacuum pump separately – without having to disconnect it from the system
- Choice of 3 operation modes (vacufuge, centrifuge, and desiccator) for a variety of application needs
- Choice of three application modes (aqueous, alcohol or high vapor pressure) to correspond with sample solvent—reduction of processing time up to 20%
- Choice of 4 heating levels (ambient, 30C, 45°C, 60°C) allows safe and efficient concentration of various samples
- Brake function can optionally be turned off for use with delicate samples
Specifications
Concentrator Plus Centrifuge |