Learn More
Motic™ Condenser, Achromatic, Swing-out, BA400/BA410
Click to view available options
For Use With (Equipment):
BA400 and BA410 Series Upright Compound Microscopes
Unit Size:
Each
Description
- Swing-out design.
- Achromatic for spherical and chromatic aberration correction.
- Includes iris diaphragm.
Specifications
Specifications
| Type | Condenser |
| Description | NA: 0.9/0.13 |
| For Use With (Equipment) | BA400 and BA410 Series Upright Compound Microscopes |
| Includes | Iris diaphragm |