Learn More
Lenz Laborglasintrumente Condensers with Receiver Acc. To Twisselmann
Product Code. 10070251
Change view
Click to view available options
Standard Taper Joint:
NS 29/32
NS 34/35
NS 45/40
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint unitSize
10070251 NS 45/40 Each
10161711 NS 29/32 Each
10643242 NS 34/35 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10070251 Supplier Lenz Laborglasintrumente Supplier No. 05420106
  • ONLINE EXCLUSIVE PRICE
    760.00 €
    501.33 € / Pack of 1
    Save 258.67 €
    34% Off
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Condensers with receiver
  • Acc. to Twisselmann
  • Made of DURAN™ tubing
Specifications
Capacity (Metric) 100 to 250 mL
Material Borosilicate Glass 3.3
Standard Taper Joint NS 45/40