missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Conditioning Reagent, for 3500/SeqStudio™ Flex

Product Code. 15839722 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 unit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15839722 1 unit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15839722 Supplier Applied Biosystems™ Supplier No. 4393718
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Conditioning Reagent is offered here as ready-to-use pouch compatible with 3500 and SeqStudio Flex series genetic analyzers. It is designed to be used for priming the polymer pump, washing the polymer pump between polymer type changes, and during instrument shut down.

Conditioning Reagent is offered here as ready-to-use pouch compatible with 3500 and SeqStudio Flex series genetic analyzers. It is designed to be used for priming the polymer pump, washing the polymer pump between polymer type changes, and during instrument shut down. The pouch has adequate volume for a one-time use. Incorporated into the label is a radio frequency identification (RFID) tag. The top part of the pouch fitment is sealed with a plastic film, which should be removed prior to installation on the instrument.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Starting Material DNA
Quantity 1 unit
Format Pouch
For Use With (Equipment) 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers
Product Type Conditioning Reagent
Shipping Condition Wet Ice
Content And Storage Store pouches at 2 to 8°C.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.