missing translation for 'onlineSavingsMsg'
Learn More

Consort™ Multi-parameter Analyzer Kits

Product Code. 11965458
Change view
Click to view available options
Type:
pH/Conductivity/DO kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11965458 pH/Conductivity/DO kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11965458 Supplier Consort™ Supplier No. C5020T

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes all the components needed for electrochemical measurement. Consort™ Multi-parameter Analyzer Kits include a high-quality, portable meter as well as the necessary electrodes; buffers, solutions and standards; and adapters and accessories required for field and laboratory use.

C5020T: pH/EC/DO Meter Kit

The C5020 is a complete and accurate portable meter that can measure all common electrochemical parameters such as pH, ORP, conductivity, and dissolved oxygen, plus salinity and TDS. It is an all-round instrument with a large conductivity range and measurement storing capabilities. The large graphical display makes it easy to read the measurement, and the stability indicator/algorithm ensures steady, accurate readings.

  • C5020 Multi-Parameter Analyzer
  • Electrodes: pH/ATC electrode SP10T, Conductivity/ATC electrode SK10T, Dissolved oxygen electrode SZ10T
  • 2 x 50mL pH buffers (pH 4 and 7)
  • 50mL Electrolyte solution (3M KCl)
  • 50mL conductivity standard (0.01M KCl)
  • Mains adaptor (100–240 VAC, EU/US)

C6010K: EC Meter Kit

The C6010 is a single channel multi-parameter analyzer suitable as a bench-top or portable device. A corrosion-resistant gold-plated BNC connector ensures a long life and accepts different electrodes. An easy-to-use menu and built-in help system enables users to configure and utilize the instrument successfully. Besides the pre-programmed pH buffers and EC standards, other tables can be added, including the complete temperature table.

  • C6010 Multi-Parameter Analyzer
  • Glass conductivity/ATC electrode SK20T
  • 50mL conductivity standard (0.01M KCl)
  • Carrying case
  • Mains adaptor (100–240 VAC, EU/US)

Specifications

Accuracy (Conductivity) 1%
Accuracy (Temperature) 0.5°C
Calibration GLP
Type pH/Conductivity/DO kit
Dimensions (L x W x H) 13 x 18 x 10 cm
Display Type LCD
Frequency 50/60Hz
Inputs BNC (pH, mV, conductivity, DO), Banana for Pt1000 (temperature)
Includes C5020 Meter, pH/ATC electrode SP10T, conductivity/ATC electrode SK10T, DO electrode SZ10T, pH buffers, electrolyte, conductivity standard, mains adapter
Model C5020T
Probe Type pH/ATC electrode SP10T, conductivity/ATC electrode SK10T, dissolved oxygen electrode SZ10T
Range (Conductivity) 0 to 1000 mS/cm
Range (Salinity) 0 to 70
Range (TDS) 0 to 100.0 g/L
Resolution (Conductivity) 0.01 μS/cm
Resolution (pH) 0.01 pH
Resolution (Temperature) 0.1°C
Resolution (TDS) 0.1 mg/L
Temperature (Metric) Range 0°C to 100°C
Voltage 100/240VAC
Weight (Metric) 600 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.