Learn More
Tuttnauer Conductivity Probe
Product Code. 17848249 Shop All Tuttnauer Products
Click to view available options
:
Conductivity Probe; For use with: Tiva Washer
Product Code. 17848249
Brand: Tuttnauer 119904380
Only null left
Add to basket
Add to basket
Conductivity Probe checks the water purity level at the end of rinsing. The machine repeats the rinse cycle again to remove any residue if the conductivity probe detects a water purity level, expressed in μS/cm, higher than the preset one.
  • Conductivity Probe Accessory
  • Checks the water purity level at the end of rinsing.
  • The machine repeats the rinse cycle again to remove any residue if the conductivity probe detects a water purity level, expressed in μS/cm, higher than the preset one.
Specifications
Material Plastic
Product Type Probe
For Use With (Equipment) Tiva Washer
Quantity 1 Ea.