missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Conduritol B epoxide

Product Code. 18788696
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18788696 5 mg 5mg
18718546 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18788696 Supplier R&D Systems™ Supplier No. 7056/5
  • ONLINE EXCLUSIVE PRICE
    166.77 €
    149.10 € / 5mg
    Save 17.67 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Irreversible glucocerebrosidase (β-glucosidase) inhibitor; used to generate Gaucher disease models

Conduritol B epoxide is an irreversible glucocerebrosidase (GBA) inhibitor (Ki = 53 μM; IC50 = 4.28 - 9.49 μM). Also inhibitis lysosomal α-glucosidase. Used to generate animal models of Gaucher disease. Accelerates recovery of peripheral nerve injury in mice. Preserves motoneurons and ganglioside distribution at the neuromuscular junction, delays disease onset and improves motor function in a mouse ALS model.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-1,2-Anhydro-myo-Inositol
CAS 6090-95-5
Quantity 5 mg
Product Line Tocris
Target Glycosylase Inhibitors
Molecular Formula C6H10O5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.