missing translation for 'onlineSavingsMsg'
Learn More

Conduritol B epoxide, MedChemExpress

Product Code. 16485947
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485947 5 mg 5mg
16473218 10 mg 10mg
16469766 50 mg 50mg
16462676 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485947 Supplier Medchem Express Supplier No. HY100944
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Conduritol B epoxide is an irreversible covalently bound acid β-glucosidase (GCase) inhibitor.

Conduritol B epoxide is an irreversible covalently bound acid β-glucosidase (GCase) inhibitor.

Specifications

Chemical Name or Material Conduritol B epoxide
CAS 6090-95-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C6H10O5
Quantity 5 mg
Solubility Information H2O : 100 mg/mL (616.75 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (308.38 mM; Need ultrasonic)
SMILES O[C@@H]1[C@@H](O)[C@H](O)[C@@H](O)[C@H]2[C@@H]1O2
Molecular Weight (g/mol) 162.14
Formula Weight 162.14
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.