missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Congo Red

Product Code. 16452316
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
Product Code. Quantity unitSize
16452316 500 mg 500mg
16432419 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 16452316

Brand: MedChemExpress HYD0236

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Congo Red is an azo dye. Congo Red (CR) binding been used as a diagnostic test for the presence of amyloid in tissue sections.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Product Type Dye Reagents
Quantity 500 mg
Shipping Condition Room temperature
CAS 573-58-0
Molecular Weight (g/mol) 696.66 g/mol
Target HBV
Solubility DMSO : 20 mg/mL (28.71 mM; Need ultrasonic) ∣ H2O : < 0.1 mg/mL (insoluble)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.